Assay Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020603.2 | NP_065628.1 | 4 - 5 | 484 | 88 | |
| GenBank mRNA | AK216977.1 | - | 2 - 3 | 145 | 88 | |
| BC013527.1 | - | 4 - 5 | 527 | 88 | ||
| BC026431.1 | - | 1 - 2 | 108 | 88 | ||
| BC046977.1 | - | 4 - 5 | 484 | 6315172 | 88 | |
| KJ650211.1 | - | 4 - 5 | 469 | 88 | ||
| KJ663716.1 | - | 4 - 5 | 469 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wdr46
Genomic Map