Assay Details
| Gene Symbol: Tap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013683.2 | NP_038711.2 | 5 - 6 | 1498 | 103 | |
| GenBank mRNA | AK040919.1 | - | 3 - 4 | 2461 | 103 | |
| AK151481.1 | - | 5 - 6 | 1253 | 103 | ||
| AK152953.1 | - | 5 - 6 | 1198 | 103 | ||
| AK153890.1 | - | 5 - 6 | 1502 | 103 | ||
| AK162731.1 | - | 5 - 6 | 1298 | 103 | ||
| AK166046.1 | - | 5 - 6 | 1499 | 103 | ||
| AK167485.1 | - | 5 - 6 | 1500 | 103 | ||
| BC013802.1 | - | 5 - 6 | 909 | 3257566 | 103 | |
| BC024897.1 | - | 5 - 6 | 1021 | 3257734 | 103 | |
| BC031584.1 | - | 1 - 2 | 1633 | 103 | ||
| KJ650227.1 | - | 5 - 6 | 1174 | 103 | ||
| M55637.1 | - | 5 - 6 | 890 | 103 | ||
| U60018.1 | - | 5 - 6 | 1174 | 103 | ||
| U60019.1 | - | 5 - 6 | 1174 | 103 | ||
| U60020.1 | - | 5 - 6 | 1174 | 103 | ||
| U60021.1 | - | 5 - 6 | 1174 | 103 | ||
| U60022.1 | - | 5 - 6 | 1174 | 103 | ||
| U60023.1 | - | 5 - 6 | 1174 | 103 | ||
| X59615.1 | - | 5 - 6 | 733 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tap1
Genomic Map