Assay Details
| Gene Symbol: Adgrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289499.1 | NP_001276428.1 | 7 - 8 | 2160 | 112 | |
| NM_001289500.1 | NP_001276429.1 | 7 - 8 | 2147 | 112 | ||
| NM_001289501.1 | NP_001276430.1 | 7 - 8 | 2191 | 112 | ||
| NM_030067.2 | NP_084343.1 | 7 - 8 | 2234 | 112 | ||
| GenBank mRNA | AK019508.1 | - | 7 - 8 | 2237 | 112 | |
| BC089564.1 | - | 7 - 8 | 2160 | 30502915 | 112 | |
| BC140980.1 | - | 7 - 8 | 2100 | 9055592 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987146 | AK019508.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987146 | NM_030067.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913884 | AF125570.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913884 | AF125571.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913884 | AF125572.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913884 | AK007663.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913884 | BC028505.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913884 | NM_011332.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725373 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211764 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526292 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838207 | AK140533.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838207 | AK142198.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838207 | NM_001039373.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838207 | NM_010839.5 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838207 | U32332.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838207 | Z35294.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15435 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20944 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46881 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142480 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179528 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987146 | AK019508.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987146 | NM_030067.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08780385 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08780435 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142480 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179528 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08780452 | - | - |
| Whole Mouse Genome G4122A | A_51_P304977 | AK019508.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P304977 | BC089564.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P304977 | NM_030067.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179528 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13297925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13082524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13157681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13157676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13306514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13297934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13398324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13398328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13082521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13430238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13133096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13207780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13182414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13549983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31185424 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450988 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429460_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450982 | - | - |
| Mouse Genome 430 2.0 Array | 1429460_at | - | - |
| Mouse Genome 430 2.0 Array | 1429460_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78249_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78249_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019787.mm.1 | NM_001289499.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019787.mm.1 | NM_001289500.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019787.mm.1 | NM_001289501.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019787.mm.1 | NM_030067.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019787.mm.1 | TC1700002114.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019788.mm.1 | NM_001289499.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019788.mm.1 | NM_001289500.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019788.mm.1 | NM_001289501.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019788.mm.1 | NM_030067.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019788.mm.1 | TC1700002114.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019789.mm.1 | NM_001289499.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019789.mm.1 | NM_001289500.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019789.mm.1 | NM_001289501.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019789.mm.1 | NM_030067.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019789.mm.1 | TC1700002114.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019786.mm.1 | NM_001289499.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019786.mm.1 | NM_001289500.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019786.mm.1 | NM_001289501.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019786.mm.1 | NM_030067.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019786.mm.1 | TC1700002114.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429460_PM_at | - | - |
| Mouse Expression Set 430 | 1429460_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429460_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429460_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 580113 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2360341 | - | - |

Adgrf4
Genomic Map