Assay Details
| Gene Symbol: Adgrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289499.1 | NP_001276428.1 | 8 - 9 | 2214 | 103 | |
| NM_001289500.1 | NP_001276429.1 | 8 - 9 | 2201 | 103 | ||
| NM_001289501.1 | NP_001276430.1 | 8 - 9 | 2245 | 103 | ||
| NM_030067.2 | NP_084343.1 | 8 - 9 | 2288 | 103 | ||
| GenBank mRNA | AK019508.1 | - | 8 - 9 | 2291 | 103 | |
| BC140980.1 | - | 8 - 9 | 2154 | 9055592 | 103 | |
More Information
Set Membership: |
|

Adgrf4
Genomic Map