Assay Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025989.3 | NP_080265.2 | 1 - 2 | 86 | 104 | |
| GenBank mRNA | AK009661.1 | - | 1 - 2 | 87 | 104 | |
| BC119573.2 | - | 1 - 2 | 40057995 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009661.1, BC119573.2, NM_025989.3 transcripts.
Set Membership: |
|

Gp2
Genomic Map