Assay Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025989.3 | NP_080265.2 | 9 - 10 | 1638 | 64 | |
| GenBank mRNA | AK009466.1 | - | 4 - 5 | 535 | 64 | |
| AK009558.1 | - | 3 - 4 | 348 | 64 | ||
| AK009661.1 | - | 9 - 10 | 1639 | 64 | ||
| BC119573.2 | - | 9 - 10 | 1633 | 40057995 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gp2
Genomic Map