| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200013048.mm.1 |
NM_001045523.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013048.mm.1 |
NM_001045523.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013048.mm.1 |
TC0200001750.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200013047.mm.1 |
NM_001045523.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013047.mm.1 |
NM_001045523.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013047.mm.1 |
TC0200001750.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200013049.mm.1 |
NM_001045523.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013049.mm.1 |
NM_001045523.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013049.mm.1 |
TC0200001750.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200013045.mm.1 |
NM_001045523.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013045.mm.1 |
NM_001045523.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200013045.mm.1 |
TC0200001750.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200013044.mm.1 |
TC0200001750.mm.1 |
MTA TC |