Assay Details
| Gene Symbol: Bahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045523.1 | NP_001038988.1 | 3 - 4 | 1946 | 69 | |
| GenBank mRNA | AK129243.1 | - | 3 - 4 | 2104 | 69 | |
| AK152597.1 | - | 3 - 4 | 1885 | 69 | ||
| BC047433.1 | - | 2 - 3 | 695 | 69 | ||
| BC100358.1 | - | 3 - 4 | 2108 | 30848279 | 69 | |
| BC141046.1 | - | 2 - 3 | 1809 | 9055658 | 69 | |
| BC145233.1 | - | 2 - 3 | 1809 | 9053785 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bahd1
Genomic Map