Assay Details
| Gene Symbol: Bahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045523.1 | NP_001038988.1 | 5 - 6 | 2181 | 93 | |
| GenBank mRNA | AK129243.1 | - | 5 - 6 | 2339 | 93 | |
| AK152597.1 | - | 5 - 6 | 2120 | 93 | ||
| BC047433.1 | - | 4 - 5 | 930 | 93 | ||
| BC100358.1 | - | 5 - 6 | 2343 | 30848279 | 93 | |
| BC141046.1 | - | 4 - 5 | 2044 | 9055658 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bahd1
Genomic Map