Assay Details
| Gene Symbol: Adm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009627.1 | NP_033757.1 | 1 - 2 | 150 | 106 | |
| GenBank mRNA | AK144181.1 | - | 1 - 2 | 165 | 106 | |
| AK144600.1 | - | 1 - 2 | 159 | 106 | ||
| AK144649.1 | - | 1 - 2 | 163 | 106 | ||
| AK171066.1 | - | 1 - 2 | 159 | 106 | ||
| BC052665.1 | - | 1 - 2 | 160 | 30054008 | 106 | |
| U77630.1 | - | 1 - 2 | 150 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144181.1, AK144600.1, AK144649.1, AK171066.1, BC052665.1, NM_009627.1, U77630.1 transcripts.
Set Membership: |
|

Adm
Genomic Map