Assay Details
| Gene Symbol: 4833423E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081664.2 | NP_001075133.1 | 8 - 9 | 1278 | 77 | |
| GenBank mRNA | AK076485.1 | - | 8 - 9 | 1367 | 77 | |
| BC120879.2 | - | 8 - 9 | 1278 | 40057526 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

4833423E24Rik
Genomic Map