Assay Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144818.3 | NP_659067.2 | 13 - 14 | 1765 | 88 | |
| GenBank mRNA | AK028919.1 | - | 13 - 14 | 1750 | 88 | |
| AK042706.1 | - | 13 - 14 | 1682 | 88 | ||
| AK045239.1 | - | 1 - 2 | 1115 | 88 | ||
| AK080433.1 | - | 13 - 14 | 1754 | 88 | ||
| AK129047.1 | - | 10 - 11 | 1318 | 88 | ||
| BC021499.1 | - | 13 - 14 | 1739 | 5346298 | 88 | |
| BC025060.1 | - | 13 - 14 | 1690 | 3673890 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncaph
Genomic Map