Assay Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144818.3 | NP_659067.2 | 6 - 7 | 788 | 95 | |
| GenBank mRNA | AK028919.1 | - | 6 - 7 | 773 | 95 | |
| AK042706.1 | - | 6 - 7 | 705 | 95 | ||
| AK080433.1 | - | 6 - 7 | 777 | 95 | ||
| AK129047.1 | - | 3 - 4 | 341 | 95 | ||
| BC021499.1 | - | 6 - 7 | 762 | 5346298 | 95 | |
| BC025060.1 | - | 6 - 7 | 713 | 3673890 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncaph
Genomic Map