| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200013325.mm.1 |
NM_010157.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013325.mm.1 |
NM_010157.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013325.mm.1 |
NM_207707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013325.mm.1 |
NM_207707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013325.mm.1 |
NR_104386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013325.mm.1 |
TC1200001942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013324.mm.1 |
NM_010157.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013324.mm.1 |
NM_010157.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013324.mm.1 |
NM_207707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013324.mm.1 |
NM_207707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013324.mm.1 |
NR_104386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013324.mm.1 |
TC1200001942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013326.mm.1 |
NM_010157.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013326.mm.1 |
NM_010157.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013326.mm.1 |
NM_207707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013326.mm.1 |
NM_207707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013326.mm.1 |
NR_104386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013326.mm.1 |
TC1200001942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013323.mm.1 |
NM_010157.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013323.mm.1 |
NM_010157.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013323.mm.1 |
NM_207707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013323.mm.1 |
NM_207707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013323.mm.1 |
NR_104386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013323.mm.1 |
TC1200001942.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200013329.mm.1 |
NM_010157.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013329.mm.1 |
NM_010157.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013329.mm.1 |
NM_207707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013329.mm.1 |
NM_207707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013329.mm.1 |
NR_104386.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200013329.mm.1 |
TC1200001942.mm.1 |
MTA TC |