Assay Details
| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010633.3 | NP_034763.3 | 6 - 7 | 1177 | 97 | |
| GenBank mRNA | AK013347.1 | - | 6 - 7 | 1182 | 97 | |
| AK030152.1 | - | 994 | 97 | |||
| BC058732.1 | - | 6 - 7 | 1177 | 6414877 | 97 | |
| Y10725.2 | - | 6 - 7 | 1034 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uhmk1
Genomic Map