Assay Details
| Gene Symbol: Fate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003243.2 | 1 - 2 | 71 | 95 | ||
| GenBank mRNA | AK020359.1 | - | 1 - 2 | 72 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline rich Gla (G-carboxyglutamic acid) 3 (transmembrane) | Prrg3 | Gm368 | -264.0 | Sense | Chr. X - 71962156 - 71972722 on NCBI Build GRCm38 |
| Downstream | cyclic nucleotide gated channel alpha 2 | Cnga2 |
CNG2, Cnca, Cncg4, OCNC1 |
8.6kb | Sense | Chr. X - 71991875 - 72010218 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK020359.1 transcripts.
Set Membership: |
|

Fate1
Genomic Map