Assay Details
| Gene Symbol: Hspa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008300.3 | NP_032326.3 | 2 - 3 | 478 | 113 | |
| GenBank mRNA | AI316935.1 | - | 2 - 3 | 478 | 113 | |
| AK053601.1 | - | 2 - 3 | 411 | 113 | ||
| AK054010.1 | - | 2 - 3 | 419 | 113 | ||
| AK133772.1 | - | 2 - 3 | 419 | 113 | ||
| AK155305.1 | - | 2 - 3 | 395 | 113 | ||
| AK166102.1 | - | 2 - 3 | 463 | 113 | ||
| AK183573.1 | - | 1 - 2 | 59 | 113 | ||
| AK208646.1 | - | 1 - 2 | 78 | 113 | ||
| AK220167.1 | - | 2 - 3 | 429 | 113 | ||
| BC003770.1 | - | 2 - 3 | 387 | 3494648 | 113 | |
| BC003950.1 | - | 2 - 3 | 373 | 113 | ||
| D85904.1 | - | 2 - 3 | 180 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa4
Genomic Map