Assay Details
| Gene Symbol: Hspa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008300.3 | NP_032326.3 | 3 - 4 | 613 | 135 | |
| GenBank mRNA | AK054010.1 | - | 3 - 4 | 554 | 135 | |
| AK133772.1 | - | 3 - 4 | 554 | 135 | ||
| AK155305.1 | - | 3 - 4 | 530 | 135 | ||
| AK166102.1 | - | 3 - 4 | 598 | 135 | ||
| AK183573.1 | - | 2 - 3 | 194 | 135 | ||
| AK220167.1 | - | 3 - 4 | 564 | 135 | ||
| BC003770.1 | - | 3 - 4 | 522 | 3494648 | 135 | |
| BC003950.1 | - | 3 - 4 | 508 | 135 | ||
| D85904.1 | - | 3 - 4 | 315 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa4
Genomic Map