Assay Details
| Gene Symbol: Hspa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008300.3 | NP_032326.3 | 6 - 7 | 981 | 93 | |
| GenBank mRNA | AK053601.1 | - | 6 - 7 | 915 | 93 | |
| AK054010.1 | - | 6 - 7 | 922 | 93 | ||
| AK133772.1 | - | 6 - 7 | 923 | 93 | ||
| AK155305.1 | - | 6 - 7 | 898 | 93 | ||
| AK166102.1 | - | 6 - 7 | 966 | 93 | ||
| AK220167.1 | - | 6 - 7 | 932 | 93 | ||
| BC003770.1 | - | 6 - 7 | 890 | 3494648 | 93 | |
| BC003950.1 | - | 6 - 7 | 876 | 93 | ||
| CF741432.1 | - | 4 - 5 | 400 | 93 | ||
| D85904.1 | - | 6 - 7 | 683 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa4
Genomic Map