Assay Details
| Gene Symbol: Mtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164190.1 | NP_001157662.1 | 5 - 6 | 539 | 89 | |
| NM_001164191.1 | NP_001157663.1 | 5 - 6 | 430 | 89 | ||
| NM_001164192.1 | NP_001157664.1 | 5 - 6 | 430 | 89 | ||
| NM_001164193.1 | NP_001157665.1 | 5 - 6 | 430 | 89 | ||
| NM_019926.3 | NP_064310.2 | 5 - 6 | 419 | 89 | ||
| GenBank mRNA | AK016445.1 | - | 5 - 6 | 540 | 89 | |
| AK031794.1 | - | 5 - 6 | 401 | 89 | ||
| AK149997.1 | - | 5 - 6 | 432 | 89 | ||
| AK166452.1 | - | 5 - 6 | 398 | 89 | ||
| AK171475.1 | - | 5 - 6 | 408 | 89 | ||
| BC051022.1 | - | 5 - 6 | 388 | 89 | ||
| BC066847.1 | - | 5 - 6 | 377 | 89 | ||
| BC090984.1 | - | 5 - 6 | 397 | 30246206 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtm1
Genomic Map