Assay Details
| Gene Symbol: Atg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025770.3 | NP_080046.3 | 1 - 2 | 49 | 62 | |
| GenBank mRNA | AK017355.1 | - | 1 - 2 | 31 | 62 | |
| BB843097.1 | - | 1 - 2 | 49 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017355.1, NM_025770.3 transcripts.
Set Membership: |
|

Atg10
Genomic Map