Assay Details
| Gene Symbol: Gabrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290435.1 | NP_001277364.1 | 1 - 2 | 420 | 61 | |
| NM_020488.2 | NP_065234.1 | 1 - 2 | 420 | 61 | ||
| GenBank mRNA | AF189260.1 | - | 1 - 2 | 162 | 61 | |
| AK038859.1 | - | 1 - 2 | 331 | 61 | ||
| BC119074.1 | - | 1 - 2 | 160 | 8733823 | 61 | |
| BC119076.1 | - | 1 - 2 | 160 | 8733825 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrq
Genomic Map