| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300007048.mm.1 |
NM_007930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007048.mm.1 |
NM_007930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007048.mm.1 |
TC1300001100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007045.mm.1 |
NM_007930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007045.mm.1 |
NM_007930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007045.mm.1 |
TC1300001100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007047.mm.1 |
NM_007930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007047.mm.1 |
NM_007930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007047.mm.1 |
TC1300001100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007046.mm.1 |
NM_007930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007046.mm.1 |
NM_007930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007046.mm.1 |
TC1300001100.mm.1 |
MTA TC |