Assay Details
| Gene Symbol: Hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010422.2 | NP_034552.1 | 6 - 7 | 801 | 96 | |
| GenBank mRNA | AK151149.1 | - | 6 - 7 | 738 | 96 | |
| AK159134.1 | - | 6 - 7 | 754 | 96 | ||
| AK159178.1 | - | 6 - 7 | 775 | 96 | ||
| AK160560.1 | - | 6 - 7 | 770 | 96 | ||
| AK168825.1 | - | 6 - 7 | 775 | 96 | ||
| AK209131.1 | - | 3 - 4 | 248 | 96 | ||
| U07633.1 | - | 6 - 7 | 773 | 96 | ||
| Y00964.1 | - | 6 - 7 | 714 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexb
Genomic Map