Assay Details
| Gene Symbol: Gcnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166065.1 | NP_001159537.1 | 2 - 3 | 378 | 99 | |
| GenBank mRNA | AK157788.1 | - | 2 - 3 | 379 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001166065.1 transcripts.
Set Membership: |
|

Gcnt4
Genomic Map