| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300017295.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017295.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017295.mm.1 |
TC1300002540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017293.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017293.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017293.mm.1 |
TC1300002540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017294.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017294.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017294.mm.1 |
TC1300002540.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300017297.mm.1 |
NM_009874.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017297.mm.1 |
NM_009874.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300017297.mm.1 |
TC1300002540.mm.1 |
MTA TC |