Assay Details
| Gene Symbol: Ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008756.2 | NP_032782.1 | 4 - 5 | 1058 | 128 | |
| GenBank mRNA | AK019880.1 | - | 4 - 5 | 1010 | 128 | |
| AK033532.1 | - | 4 - 5 | 991 | 128 | ||
| AK042576.1 | - | 4 - 5 | 1058 | 128 | ||
| AK145798.1 | - | 2 - 3 | 217 | 128 | ||
| BC138679.1 | - | 4 - 5 | 1041 | 8861700 | 128 | |
| BC138680.1 | - | 4 - 5 | 1041 | 8861701 | 128 | |
| BC145371.1 | - | 4 - 5 | 1056 | 9053929 | 128 | |
| U49185.1 | - | 4 - 5 | 1101 | 128 | ||
More Information
Set Membership: |
|

Ocln
Genomic Map