Assay Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144654.1 | - | 11 - 12 | 1515 | 75 | |
| BC027130.1 | - | 1 - 2 | 126 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdm6a| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144654.1 | - | 11 - 12 | 1515 | 75 | |
| BC027130.1 | - | 1 - 2 | 126 | 75 | ||
Set Membership: |
|