Assay Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009483.1 | NP_033509.1 | 14 - 15 | 1764 | 82 | |
| GenBank mRNA | AJ002730.1 | - | 13 - 14 | 1223 | 82 | |
| AK080370.1 | - | 14 - 15 | 1783 | 82 | ||
| AK144654.1 | - | 12 - 13 | 1600 | 82 | ||
| BC027130.1 | - | 2 - 3 | 211 | 82 | ||
| BC053433.1 | - | 14 - 15 | 1764 | 6516368 | 82 | |
| BC075703.1 | - | 5 - 6 | 674 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kdm6a
Genomic Map