Assay Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009483.1 | NP_033509.1 | 20 - 21 | 3481 | 72 | |
| GenBank mRNA | AJ002730.1 | - | 19 - 20 | 2940 | 72 | |
| AK144654.1 | - | 18 - 19 | 3316 | 72 | ||
| AK162600.1 | - | 4 - 5 | 765 | 72 | ||
| AK164248.1 | - | 1 - 2 | 98 | 72 | ||
| BC027130.1 | - | 8 - 9 | 1928 | 72 | ||
| BC053433.1 | - | 20 - 21 | 3481 | 6516368 | 72 | |
| BC075703.1 | - | 11 - 12 | 2391 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kdm6a
Genomic Map