| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016886.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016886.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016886.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016886.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016886.mm.1 |
TC1300002510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016888.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016888.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016888.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016888.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016888.mm.1 |
TC1300002510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016889.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016889.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016889.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016889.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016889.mm.1 |
TC1300002510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016887.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016887.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016887.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016887.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016887.mm.1 |
TC1300002510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016890.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016890.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016890.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016890.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016890.mm.1 |
TC1300002510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016884.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016884.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016884.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016884.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016884.mm.1 |
TC1300002510.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016885.mm.1 |
NM_001048267.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016885.mm.1 |
NM_001048267.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016885.mm.1 |
NM_178716.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016885.mm.1 |
NM_178716.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016885.mm.1 |
TC1300002510.mm.1 |
MTA TC |