Assay Details
| Gene Symbol: Igsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162884.1 | NP_001156356.1 | 5 - 6 | 5550 | 82 | |
| GenBank mRNA | AK081990.1 | - | 1 - 2 | 1095 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf10
Genomic Map