Assay Details
| Gene Symbol: Igsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162884.1 | NP_001156356.1 | 6 - 7 | 6440 | 79 | |
| GenBank mRNA | AK081990.1 | - | 2 - 3 | 1986 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157897 | AK129412.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157897 | NM_177855.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157897 | NM_001162884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157902 | AK132367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2157902 | NM_001162884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155843 | AK081990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P155843 | AK081990.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729481 | AK005806.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862567 | BC019190.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862567 | NM_007986.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862567 | Y10007.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267784 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37805 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141482 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175018 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174324 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117259 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155843 | AK081990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P155843 | AK081990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157902 | AK132367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157902 | NM_001162884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157897 | AK129412.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157897 | NM_177855.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2157897 | NM_001162884.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174324 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141482 | - | - |
| Whole Mouse Genome G4122A | A_52_P429964 | AK040638.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155843 | AK081990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155843 | AK081990.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141482 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02851765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02851759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02589087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22173414 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10498390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498381 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498380 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498382 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498385 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443646_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498383 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498379 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498386 | - | - |
| Mouse Genome 430 2.0 Array | 1443646_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443646_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300015530.mm.1 | NM_001162884.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015530.mm.1 | NM_001162884.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015530.mm.1 | TC0300002056.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443646_at | - | - |
| Mouse Expression Set 430 | 1443646_PM_at | - | - |
| Murine Genome U74v2 Set | 104435_at | - | - |
| Murine Genome U74v2 Set | 104435_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443646_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443646_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 715042 | - | - |
| ABI Mouse Genome Survey Microarray | 742598 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105220047 | - | - |
| MouseWG-6 v2 Expression BeadChip | 105550086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244809 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2445658 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244809 | - | - |

Igsf10
Genomic Map