Assay Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009012.2 | NP_033038.2 | 18 - 19 | 3177 | 158 | |
| GenBank mRNA | AK037641.1 | - | 17 - 18 | 3020 | 158 | |
| U66887.1 | - | 18 - 19 | 3155 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad50
Genomic Map