Assay Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009012.2 | NP_033038.2 | 20 - 21 | 3432 | 98 | |
| GenBank mRNA | U66887.1 | - | 20 - 21 | 3410 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad50
Genomic Map