Assay Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009012.2 | NP_033038.2 | 4 - 5 | 815 | 87 | |
| GenBank mRNA | AK018001.1 | - | 4 - 5 | 815 | 87 | |
| AK037641.1 | - | 4 - 5 | 816 | 87 | ||
| BC058180.1 | - | 4 - 5 | 791 | 87 | ||
| U66887.1 | - | 4 - 5 | 793 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad50
Genomic Map