Assay Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010431.2 | NP_034561.2 | 12 - 13 | 2553 | 77 | |
| GenBank mRNA | AK033471.1 | - | 1 - 2 | 153 | 77 | |
| AK034087.1 | - | 12 - 13 | 2519 | 77 | ||
| AK076395.1 | - | 10 - 11 | 1841 | 77 | ||
| AK150367.1 | - | 12 - 13 | 2420 | 77 | ||
| BC026139.1 | - | 12 - 13 | 2405 | 4019056 | 77 | |
| X95580.1 | - | 12 - 13 | 2177 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hif1a
Genomic Map