Assay Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010431.2 | NP_034561.2 | 2 - 3 | 653 | 80 | |
| GenBank mRNA | AF003695.1 | - | 2 - 3 | 487 | 80 | |
| AK017853.1 | - | 2 - 3 | 514 | 80 | ||
| AK034087.1 | - | 2 - 3 | 619 | 80 | ||
| AK138944.1 | - | 2 - 3 | 520 | 80 | ||
| AK150367.1 | - | 2 - 3 | 520 | 80 | ||
| BC026139.1 | - | 2 - 3 | 505 | 4019056 | 80 | |
| X95580.1 | - | 2 - 3 | 319 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hif1a
Genomic Map