Assay Details
| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019993.3 | NP_064377.2 | 8 - 9 | 1451 | 106 | |
| GenBank mRNA | AB296386.1 | - | 8 - 9 | 1396 | 106 | |
| AF170919.1 | - | 8 - 9 | 1196 | 106 | ||
| AK149589.1 | - | 8 - 9 | 1237 | 106 | ||
| AK150093.1 | - | 8 - 9 | 1194 | 106 | ||
| AK154914.1 | - | 8 - 9 | 1435 | 106 | ||
| AK162354.1 | - | 8 - 9 | 1235 | 106 | ||
| AK165730.1 | - | 8 - 9 | 1451 | 106 | ||
| AK185844.1 | - | 2 - 3 | 243 | 105 | ||
| BC003297.1 | - | 8 - 9 | 1228 | 3583976 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh9a1
Genomic Map