Assay Details
| Gene Symbol: Mipol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164370.1 | NP_001157842.1 | 3 - 4 | 518 | 179 | |
| GenBank mRNA | AK006974.1 | - | 427 | 179 | ||
| AK044766.1 | - | 2 - 3 | 245 | 179 | ||
| AK050030.1 | - | 3 - 4 | 504 | 179 | ||
| AK077915.1 | - | 3 - 4 | 498 | 179 | ||
| AK085510.1 | - | 3 - 4 | 513 | 179 | ||
| AK163490.1 | - | 3 - 4 | 498 | 179 | ||
| AK168989.1 | - | 2 - 3 | 303 | 179 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mipol1
Genomic Map