Assay Details
| Gene Symbol: Mipol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164370.1 | NP_001157842.1 | 6 - 7 | 796 | 131 | |
| GenBank mRNA | AK006974.1 | - | 705 | 131 | ||
| AK044766.1 | - | 5 - 6 | 523 | 131 | ||
| AK077915.1 | - | 6 - 7 | 776 | 131 | ||
| AK138175.1 | - | 289 | 131 | |||
| AK168989.1 | - | 5 - 6 | 581 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mipol1
Genomic Map