Assay Details
| Gene Symbol: Mipol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164370.1 | NP_001157842.1 | 8 - 9 | 1074 | 66 | |
| GenBank mRNA | AK077915.1 | - | 8 - 9 | 1054 | 66 | |
| AK138175.1 | - | 567 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Mipol1
Genomic Map