Assay Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293313.1 | NP_001280242.1 | 1 - 2 | 84 | 92 | |
| NM_138314.4 | NP_612187.2 | 1 - 2 | 84 | 92 | ||
| NM_178071.6 | NP_835172.1 | 1 - 2 | 84 | 92 | ||
| GenBank mRNA | AK043515.1 | - | 1 - 2 | 68 | 92 | |
| AK133221.1 | - | 552 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133221.1, NM_001293313.1, NM_138314.4, NM_178071.6 transcripts.
Set Membership: |
|

Nme7
Genomic Map