Assay Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293313.1 | NP_001280242.1 | 9 - 10 | 1106 | 81 | |
| NM_138314.4 | NP_612187.2 | 9 - 10 | 1109 | 81 | ||
| GenBank mRNA | AF202048.1 | - | 8 - 9 | 1010 | 81 | |
| AK044962.1 | - | 9 - 10 | 1080 | 81 | ||
| AK133221.1 | - | 1577 | 81 | |||
| AK144915.1 | - | 9 - 10 | 1110 | 81 | ||
| BC038021.1 | - | 10 - 11 | 1136 | 3963844 | 81 | |
| BC049225.1 | - | 8 - 9 | 1037 | 3483725 | 81 | |
| BU609451.1 | - | 5 - 6 | 81 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nme7
Genomic Map