Assay Details
| Gene Symbol: Dcun1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205361.1 | NP_001192290.1 | 4 - 5 | 703 | 148 | |
| NM_001205362.1 | NP_001192291.1 | 4 - 5 | 663 | 148 | ||
| NM_033623.6 | NP_296372.3 | 4 - 5 | 650 | 148 | ||
| GenBank mRNA | AF198092.1 | - | 4 - 5 | 541 | 148 | |
| AK008657.1 | - | 4 - 5 | 540 | 148 | ||
| AK139845.1 | - | 4 - 5 | 653 | 148 | ||
| AK155081.1 | - | 4 - 5 | 574 | 148 | ||
| AK165651.1 | - | 4 - 5 | 576 | 148 | ||
| AK165675.1 | - | 4 - 5 | 548 | 148 | ||
| BC020161.1 | - | 4 - 5 | 626 | 3988680 | 148 | |
| BC031666.1 | - | 4 - 5 | 559 | 4012394 | 148 | |
| BC037431.1 | - | 4 - 5 | 559 | 2648141 | 148 | |
| Gene Symbol: Tes3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029314.1 | - | 1 | 856 | 148 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcun1d1
Genomic Map