| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1456360_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1456360_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1456360_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
NM_001205361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
NM_001205361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
NM_001205362.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
NM_001205362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
NM_033623.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
NM_033623.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014207.mm.1 |
TC0300001841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
NM_001205361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
NM_001205361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
NM_001205362.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
NM_001205362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
NM_033623.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
NM_033623.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014205.mm.1 |
TC0300001841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
NM_001205361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
NM_001205361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
NM_001205362.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
NM_001205362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
NM_033623.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
NM_033623.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014204.mm.1 |
TC0300001841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
NM_001205361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
NM_001205361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
NM_001205362.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
NM_001205362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
NM_033623.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
NM_033623.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014202.mm.1 |
TC0300001841.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
NM_001205361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
NM_001205361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
NM_001205362.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
NM_001205362.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
NM_033623.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
NM_033623.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014206.mm.1 |
TC0300001841.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1456360_at |
- |
- |
| Mouse Expression Set 430 |
1456360_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1456360_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1456360_PM_at |
- |
- |