Assay Details
| Gene Symbol: Prkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008858.3 | NP_032884.2 | 11 - 12 | 1933 | 87 | |
| GenBank mRNA | Z34524.1 | - | 11 - 12 | 1808 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd1
Genomic Map