Assay Details
| Gene Symbol: Prpf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177806.3 | NP_808474.2 | 11 - 12 | 1914 | 128 | |
| GenBank mRNA | AK154170.1 | - | 12 - 13 | 2538 | 128 | |
| AK168462.1 | - | 11 - 12 | 1910 | 128 | ||
| BC029153.1 | - | 6 - 7 | 886 | 128 | ||
| BC030904.1 | - | 11 - 12 | 1853 | 128 | ||
| BC033575.1 | - | 6 - 7 | 3981 | 128 | ||
| BC034078.1 | - | 14 - 15 | 2171 | 128 | ||
| BC076622.1 | - | 11 - 12 | 1600 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf39
Genomic Map