Assay Details
| Gene Symbol: Prpf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177806.3 | NP_808474.2 | 1 - 2 | 136 | 90 | |
| GenBank mRNA | AK168462.1 | - | 1 - 2 | 129 | 90 | |
| BY069337.1 | - | 1 - 2 | 137 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_177806.3 transcripts.
Set Membership: |
|

Prpf39
Genomic Map