Assay Details
| Gene Symbol: Prpf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177806.3 | NP_808474.2 | 2 - 3 | 475 | 64 | |
| GenBank mRNA | AK154170.1 | - | 2 - 3 | 476 | 64 | |
| AK168462.1 | - | 2 - 3 | 468 | 64 | ||
| BC034078.1 | - | 2 - 3 | 454 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154170.1 transcripts.
Set Membership: |
|

Prpf39
Genomic Map